3,4-Dichloro-D-phenylalanine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (R)-2-Amino-3-(3,4-dichlorophenyl)propanoic acid, H-3,4-Dichloro-D-Phe-OH, H-D-Phe(3,4-DiCl)-OH, 2-Amino-3-3-4-dichlorophenyl-propanoic-acid, D-3,4-Dichlorophenylalanine

OR14720-10G OR14720-25G OR14720-5G
77405-090EA 195.62 USD
77405-090 77405-092 77405-094
3,4-Dichloro-D-phenylalanine
3,4-Dichloro-D-phenylalanine
Formula: C₉H₉Cl₂NO₂
MW: 234.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671682
CAS Number: 52794-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR