(3-Carbamoylphenyl)boronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR10326-100G OR10326-25G OR10326-5G
77402-588EA 188.48 USD
77402-588 77402-590 77402-592
(3-Carbamoylphenyl)boronic acid 98%
(3-Carbamoylphenyl)boronic acid
Formula: C₇H₈BNO₃
Melting Pt: 232…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411948
CAS Number: 351422-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR