N-Cbz-L-asparagine 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: Z-Asn-OH, Nalpha-Cbz-L-asparagine

OR01678-100G OR01678-25G OR01678-500G
77401-800EA 61.12 USD
77401-800 77401-802 77401-804
N-Cbz-L-asparagine 97%
N-Cbz-L-asparagine
Formula: C₁₂H₁₄N₂O₅
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00008035
CAS Number: 2304-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR