4-Ethoxy-1,1,1-trifluorobut-3-en-2-one

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2-Ethoxyvinyl) trifluoromethyl ketone, (3E)-4-ethoxy-1,1,1-trifluorobut-3-en-2-one, 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, 4-ethoxy-1,1,1-trifluorobut-3-en-2-one, 1-Ethoxy-3-oxo-4,4,4-trifluorobut-1-ene

PC31704-1G PC31704-25G PC31704-5G
77414-546EA 20.26 USD
77414-546 77414-548 77414-550
4-Ethoxy-1,1,1-trifluorobut-3-en-2-one
4-Ethoxy-1,1,1-trifluorobut-3-en-2-one
Formula: C₆H₇F₃O₂
MW: 168.115 g/mol
Boiling Pt: 51…53 °C/12 mmHg
Melting Pt: –30 °C
Density: 1.18 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192131
CAS Number: 17129-06-5
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR