4-((3-Hydroxypropyl)amino)-3-nitrophenol 95+%

Supplier: APOLLO SCIENTIFIC

OR350491-100G OR350491-25G OR350491-5G
77409-292EA 103.92 USD
77409-292 77409-294 77409-296
4-((3-Hydroxypropyl)amino)-3-nitrophenol 95+%
4-((3-Hydroxypropyl)amino)-3-nitrophenol
Formula: C₉H₁₂N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD07368637
CAS Number: 92952-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR