3-Chloro-5-methylaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR313080-5G OR313080-100G OR313080-25G
77409-034EA 21.26 USD
77409-034 77409-030 77409-032
3-Chloro-5-methylaniline 98%
3-Chloro-5-methylaniline
Formula: C7H8ClN
Storage Temperature: Freezer
MDL Number: MFCD15527197
CAS Number: 29027-20-1
UN: 2239
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR