6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC48296-25G PC48296-5G
77415-734EA 103.92 USD
77415-734 77415-736
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole 97%
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole
Formula: C₁₂H₁₃FN₂O
MW: 220.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03236724
CAS Number: 84163-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR