4-Aminoimidazole-5-carboxamide 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Amino-1H-imidazole-5-carboxamide, 5-Aminoimidazole-4-carboxamide, 5-Amino-4-imidazolecarboxamide, 5-Amino-1H-imidazole-4-carboxamide, 4-Aminoimidazole-5-carboxamide, 4-aminoimidazole-5-carboxamide, 5-Amino-3H-imidazole-4-Carboxamide

OR13642-100G OR13642-25G OR13642-5G
77404-704EA 139.58 USD
77404-704 77404-706 77404-708
4-Aminoimidazole-5-carboxamide 97%
4-Aminoimidazole-5-carboxamide
Formula: C₄H₆N₄O
MW: 126.12 g/mol
Melting Pt: 164…170 °C
Storage Temperature: Ambient
MDL Number: MFCD02181040
CAS Number: 360-97-4
EINECS: 206-641-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR