5-Methoxy-1H-pyrrolo[3,2-b]pyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR301131-1G OR301131-5G
77408-788EA 77.44 USD
77408-788 77408-790
5-Methoxy-1H-pyrrolo[3,2-b]pyridine 97%
5-Methoxy-1H-pyrrolo[3,2-b]pyridine
Formula: C8H8N2O
Melting Pt: 108…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08688601
CAS Number: 17288-40-3
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR