3-Bromo-5-(trifluoromethyl)phenylacetic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC520693-1G PC520693-5G
77416-522EA 39.74 USD
77416-522 77416-524
3-Bromo-5-(trifluoromethyl)phenylacetic acid 97%
3-Bromo-5-(trifluoromethyl)phenylacetic acid
Formula: C9H6BrF3O2
MW: 283.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194218
CAS Number: 1161362-01-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR