2,3-Dihydro-1-benzofuran-5-sulfonoylchloride

Supplier: APOLLO SCIENTIFIC
Danger

OR22992-10G OR22992-1G OR22992-500MG
77407-000EA 220.08 USD
77407-000 77407-002 77407-004
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride
Formula: C₈H₇ClO₃S
MW: 218.66 g/mol
Melting Pt: 74…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02677703
CAS Number: 115010-11-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR