2-Chloro-5-(chloromethyl)thiazole

Supplier: APOLLO SCIENTIFIC
Danger

OR4254-5G OR4254-100G OR4254-25G
77409-672EA 21.26 USD
77409-672 77409-668 77409-670
2-Chloro-5-(chloromethyl)thiazole
2-Chloro-5-(chloromethyl)thiazole
Formula: C₄H₃Cl₂NS
Melting Pt: 29…31 °C
Density: 1.503 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01073549
CAS Number: 105827-91-6
EINECS: 429-830-5
UN: 2922
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR