trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: (1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, (1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine

OR913416-10G OR913416-25G OR913416-5G
77411-678EA 48.9 USD
77411-678 77411-680 77411-682
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine 97%
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine
Formula: C₈H₁₈N₂
Storage Temperature: Ambient
MDL Number: MFCD00671528
CAS Number: 87583-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR