4-Methyl-1-piperazineacetic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR17668-10G OR17668-1G
77406-078EA 139.58 USD
77406-078 77406-080
4-Methyl-1-piperazineacetic acid 97%
4-Methyl-1-piperazineacetic acid
Formula: C₇H₁₄N₂O₂
MW: 158.1983 g/mol
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD03444554
CAS Number: 54699-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR