N-Methyltetrahydro-2H-pyran-4-amine 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Methylamino)tetrahydro-2H-pyran

OR1026-10G OR1026-1G OR1026-5G
77402-552EA 248.6 USD
77402-552 77402-554 77402-556
N-Methyltetrahydro-2H-pyran-4-amine 95%
N-Methyltetrahydro-2H-pyran-4-amine
Formula: C₆H₁₃NO
MW: 115.1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06658492
CAS Number: 220641-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR