1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: 1-[(tert-butoxy)carbonyl]azetidine-2-carboxylic acid

OR309044-1G OR309044-5G
77408-986EA 61.12 USD
77408-986 77408-988
1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 95%
1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid
Formula: C₉H₁₅NO₄
MW: 201.2197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861756
CAS Number: 159749-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR