4,6-Dichloro-2-methoxypyrimidine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR18261-100MG OR18261-1G
77406-194EA 21.26 USD
77406-194 77406-196
4,6-Dichloro-2-methoxypyrimidine 98%
4,6-Dichloro-2-methoxypyrimidine
Formula: C₅H₄Cl₂N₂O
MW: 179 g/mol
Melting Pt: 53…55 °C
Storage Temperature: Freezer
MDL Number: MFCD03428360
CAS Number: 1074-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR