4-Fluoro-2-methoxyaniline 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC49081-100G PC49081-25G
77415-844EA 99.84 USD
77415-844 77415-846
4-Fluoro-2-methoxyaniline 97%
4-Fluoro-2-methoxyaniline
Formula: C₇H₈FNO
Melting Pt: 45…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00077536
CAS Number: 450-91-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR