2,4-Dichloro-5-(trifluoromethyl)pyrimidine

Supplier: APOLLO SCIENTIFIC
Danger

PC3927-1G PC3927-5G
77415-016EA 21.26 USD
77415-016 77415-018
2,4-Dichloro-5-(trifluoromethyl)pyrimidine
2,4-Dichloro-5-(trifluoromethyl)pyrimidine
Formula: C₅HCl₂F₃N₂
MW: 216.98 g/mol
Boiling Pt: 49 °C/32 mmHg
Density: 1.609 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426408
CAS Number: 3932-97-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR