1H-Benzimidazole-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR23116-1G OR23116-25G OR23116-5G
77407-110EA 21.32 USD
77407-110 77407-112 77407-114
1H-Benzimidazole-2-carboxylic acid
1H-Benzimidazole-2-carboxylic acid
Formula: C₈H₆N₂O₂
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00721565
CAS Number: 2849-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR