Methyl-1H-pyrrole-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl Pyrrole-2-carboxylate, Methyl 1H-pyrrole-2-carboxylate, Pyrrole-2-carboxylic acid methyll ester, Methyl pyrrole-2-carboxylate

OR29375-100G OR29375-25G
77408-390EA 187.46 USD
77408-390 77408-392
Methyl-1H-pyrrole-2-carboxylate
Methyl-1H-pyrrole-2-carboxylate
Formula: C₆H₇NO₂
MW: 125.13 g/mol
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00817048
CAS Number: 1193-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR