Pyridine-3-sulfonyl chloride hydrochloride, Technical Grade
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR4393-1G
OR4393-250MG
77409-720EA
38.32
USD
77409-720
77409-722
Pyridine-3-sulfonyl chloride hydrochloride, Technical Grade
Pyridine-3-sulfonyl chloride hydrochloride
Formula:
C5H5Cl2NO2S Melting Pt: 144…145 °C Storage Temperature: Freezer |
MDL Number:
MFCD04035552 CAS Number: 42899-76-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...