Pyridine-3-sulfonyl chloride 98%

Supplier: APOLLO SCIENTIFIC

OR951579-100G OR951579-25G
77412-160EA 303.6 USD
77412-160 77412-162
Pyridine-3-sulfonyl chloride 98%
Pyridine-3-sulfonyl chloride
Formula: C5H4ClNO2S
Density: 1.46 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03452747
CAS Number: 16133-25-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR