(4-Bromophenyl)acetaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR200079-5G OR200079-1G
77406-534EA 188.46 USD
77406-534 77406-532
(4-Bromophenyl)acetaldehyde 95%
(4-Bromophenyl)acetaldehyde
Formula: C₈H₇BrO
MW: 199.0446 g/mol
Boiling Pt: 113…114 °C/7 mmHg
Melting Pt: 96…97 °C
Storage Temperature: Ambient
MDL Number: MFCD02261728
CAS Number: 27200-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR