5-Phenylthiophene-2-carboxylic acid 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
5-Phenyl-2-thiophenecarboxylic acid, 5-Phenyl-2-thenoic acid
OR23146-1G
OR23146-250MG
OR23146-5G
77407-142EA
103.92
USD
77407-142
77407-144
77407-146
5-Phenylthiophene-2-carboxylic acid 97%
5-Phenylthiophene-2-carboxylic acid
Formula:
C11H8O2S Melting Pt: 188…192 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01313432 CAS Number: 19163-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...