3-Aminopicolinic acid 97%, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Amino-2-pyridinecarboxylic acid, 3-Aminopyridine-2-carboxylic acid, 2-Pyridinecarboxylic acid, 3-amino-

OR2780T-25G OR2780T-5G
77407-990EA 158.94 USD
77407-990 77407-992
3-Aminopicolinic acid 97%, Technical Grade
3-Aminopicolinic acid
Formula: C₆H₆N₂O₂
Melting Pt: 218…222 °C
Storage Temperature: Freezer
MDL Number: MFCD00090153
CAS Number: 1462-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR