1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Pyrrolidine-3-carboxylic acid, N-BOC protected

OR3670-1G OR3670-5G
77409-344EA 21.26 USD
77409-344 77409-346
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid
Formula: C₁₀H₁₇NO₄
Melting Pt: 133…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179181
CAS Number: 59378-75-5
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR