2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

OR110960-1G OR110960-250MG
77403-596EA 168.1 USD
77403-596 77403-598
2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine
2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine
Formula: C₇H₅BrN₄
Storage Temperature: Refrigerator
MDL Number: MFCD09834926
CAS Number: 857641-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR