Morpholino(piperidin-4-yl)methanone hydrochloride 90%

Supplier: APOLLO SCIENTIFIC
Warning

OR12322-1G OR12322-250MG
77404-270EA 121.24 USD
77404-270 77404-272
Morpholino(piperidin-4-yl)methanone hydrochloride 90%
Morpholino(piperidin-4-yl)methanone hydrochloride
Formula: C₁₀H₁₉ClN₂O₂
Melting Pt: 279…286 °C
Storage Temperature: Ambient
MDL Number: MFCD05864747
CAS Number: 94467-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR