(2-Nitrophenyl)hydrazine hydrochloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Nitrophenylhydrazine hydrochloride

OR1939-100G OR1939-25G OR1939-5G
77406-454EA 79.46 USD
77406-454 77406-456 77406-458
(2-Nitrophenyl)hydrazine hydrochloride
(2-Nitrophenyl)hydrazine hydrochloride
Formula: C₆H₈ClN₃O₂
MW: 189.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00060205
CAS Number: 6293-87-4
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR