Glycidyl methyl ether 90%

Supplier: APOLLO SCIENTIFIC

Synonyms: 1,2-Epoxy-3-methoxypropane

OR947696-1G OR947696-5G
77412-130EA 21.26 USD
77412-130 77412-132
Glycidyl methyl ether 90%
Glycidyl methyl ether
Formula: C₄H₈O₂
Flash Pt: 25 °C
Storage Temperature: Freezer
MDL Number: MFCD00005140
CAS Number: 930-37-0
UN: 1992
ADR: 3+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR