3-(Trifluoromethyl)picolinic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-(Trifluoromethyl)pyridine-2-carboxylic acid, 3-(Trifluoromethy)pyridine-2-carboxylic acid

PC9130-10G PC9130-1G PC9130-5G
77418-596EA 147.72 USD
77418-596 77418-598 77418-600
3-(Trifluoromethyl)picolinic acid 97%
3-(Trifluoromethyl)picolinic acid
Formula: C₇H₄F₃NO₂
MW: 191.1100 g/mol
Melting Pt: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD07375386
CAS Number: 87407-12-3
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR