5-Cyano-2-methylpyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR29756-1G OR29756-25G OR29756-5G
77408-430EA 21.26 USD
77408-430 77408-432 77408-434
5-Cyano-2-methylpyridine 98%
5-Cyano-2-methylpyridine
Formula: C₇H₆N₂
Boiling Pt: 216…217 °C
Melting Pt: 83…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038042
CAS Number: 3222-48-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR