2-Cyano-4'-methylbiphenyl

Supplier: APOLLO SCIENTIFIC
Warning

OR10059-100G OR10059-25G OR10059-5G
77402-426EA 223.12 USD
77402-426 77402-428 77402-430
2-Cyano-4'-methylbiphenyl
2-Cyano-4'-methylbiphenyl
Formula: C₁₄H₁₁N
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00151805
CAS Number: 114772-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR