4-Cyano-1-butyne 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR70029-1G OR70029-25G OR70029-5G
77410-964EA 34.64 USD
77410-964 77410-966 77410-968
4-Cyano-1-butyne 97%
4-Cyano-1-butyne
Formula: C₅H₅N
MW: 79.999 g/mol
Boiling Pt: 79 °C/35 mmHg
Density: 0.88 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041563
CAS Number: 19596-07-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR