2-Amino-4,6-dichlorophenol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,5-Dichloro-6-hydroxyaniline

OR21268-100G OR21268-25G
77406-656EA 330.1 USD
77406-656 77406-658
2-Amino-4,6-dichlorophenol 95%
2-Amino-4,6-dichlorophenol
Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00035766
CAS Number: 527-62-8
EINECS: 208-421-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR