4,4'-Difluorobenzhydryl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Chloro bis-(4-fluorophenyl)methane, 4,4-Difluorodiphenylmethylchloride

PC4106-10G PC4106-50G PC4106-5G
77415-136EA 48.9 USD
77415-136 77415-138 77415-140
4,4'-Difluorobenzhydryl chloride 97%
4,4'-Difluorobenzhydryl chloride
Formula: C₁₃H₉ClF₂
MW: 238.66 g/mol
Boiling Pt: 126…128 °C/1 mmHg
Density: 1.28 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00044329
CAS Number: 27064-94-4
EINECS: 248-201-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR