4-Amino-8-methoxy-2-methylquinoline

Supplier: APOLLO SCIENTIFIC

OR307958-1G OR307958-250MG
77408-952EA 330.1 USD
77408-952 77408-954
4-Amino-8-methoxy-2-methylquinoline
4-Amino-8-methoxy-2-methylquinoline
Formula: C₁₁H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD08754055
CAS Number: 657391-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR