2-Sulfobenzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR61312-100G OR61312-25G OR61312-5G
77410-728EA 997.42 USD
77410-728 77410-730 77410-732
2-Sulfobenzoic acid 95%
2-Sulphobenzoic acid
Formula: C₇H₆O₅S
Melting Pt: 142 °C
Storage Temperature: Ambient
CAS Number: 632-25-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR