1-Boc-4-cyanopiperidine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: tert-Butyl 4-cyanopiperidine-1-carboxylate, N-BOC-Piperidine-4-carbonitrile

OR9060-25G OR9060-5G
77411-560EA 48.9 USD
77411-560 77411-562
1-Boc-4-cyanopiperidine 98%
1-Boc-4-cyanopiperidine
Formula: C₁₁H₁₈N₂O₂
Melting Pt: 60…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861223
CAS Number: 91419-52-2
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR