3-(Piperidin-1-ylsulfonyl)aniline 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR14248-10G OR14248-1G OR14248-5G
77404-866EA 678.54 USD
77404-866 77404-868 77404-870
3-(Piperidin-1-ylsulfonyl)aniline 97%
3-(Piperidin-1-ylsulphonyl)aniline
Formula: C₁₁H₁₆N₂O₂S
Melting Pt: 116…117 °C
Storage Temperature: Ambient
MDL Number: MFCD02063328
CAS Number: 22184-99-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR