2-(2-Naphthyl)indole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0668-1G OR0668-250MG OR0668-5G
77402-190EA 135.5 USD
77402-190 77402-192 77402-194
2-(2-Naphthyl)indole 98%
2-(2-Naphthyl)indole
Formula: C₁₈H₁₃N
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00124144
CAS Number: 23746-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR