5-Bromo-N-methylpicolinamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11592-100MG OR11592-1G OR11592-250MG
77403-964EA 21.34 USD
77403-964 77403-966 77403-968
5-Bromo-N-methylpicolinamide 98%
5-Bromo-N-methylpicolinamide
Formula: C₇H₇BrN₂O
Melting Pt: 76…77 °C
Storage Temperature: Ambient
MDL Number: MFCD07357421
CAS Number: 845305-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR