Methyl-4-methylpicolinate 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: Methyl 4-methylpicolinate

OR955702-1G OR955702-5G
77412-190EA 85.58 USD
77412-190 77412-192
Methyl-4-methylpicolinate 97%
Methyl 4-methylpicolinate
Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265118
CAS Number: 13509-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR