3-Hydroxy-5-(trifluoromethoxy)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7918-1G PC7918-500MG
77418-288EA 323.98 USD
77418-288 77418-290
3-Hydroxy-5-(trifluoromethoxy)phenylboronic acid 98%
3-Hydroxy-5-(trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₄
Melting Pt: 78…81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09475898
CAS Number: 957120-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR