3-(Methylthio)-1H-1,2,4-triazole

Supplier: APOLLO SCIENTIFIC
Warning

OR019167-1G OR019167-5G
77401-898EA 96.78 USD
77401-898 77401-900
3-(Methylthio)-1H-1,2,4-triazole
3-(Methylthio)-1H-1,2,4-triazole
Formula: C₃H₅N₃S
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD01312671
CAS Number: 7411-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR