1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine

PC0916-1G PC0916-500MG
77412-736EA 131.42 USD
77412-736 77412-738
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane 97%
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane
Formula: C₁₁H₁₃ClF₃N₃
MW: 279.69
Boiling Pt: 126…128 °C/1 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00238856
CAS Number: 231953-40-5
UN: 3267
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR