2-Bromo-4-fluoro-5-nitrophenol 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC5004-5G
PC5004-100G
PC5004-25G
77416-014EA
24
USD
77416-014
77416-006
77416-008
2-Bromo-4-fluoro-5-nitrophenol 97%
2-Bromo-4-fluoro-5-nitrophenol
Formula:
C₆H₃BrFNO₃ MW: 236.00 g/mol Melting Pt: 126…127 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08460104 CAS Number: 84478-87-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...