5-Bromo-o-toluic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR59443-25G OR59443-100G
77410-594EA 34.64 USD
77410-594 77410-592
5-Bromo-o-toluic acid
5-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 167…171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00267350
CAS Number: 79669-49-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR