Ethyl trifluoropyruvate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate

PC3288G-100G PC3288G-25G
77414-664EA 63.16 USD
77414-664 77414-666
Ethyl trifluoropyruvate 98%
Ethyl trifluoropyruvate

Useful prochiral intermediate.

Formula: C₅H₅F₃O₃
MW: 170.09
Boiling Pt: 95…98 °C
Density: 1.283 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00114935
CAS Number: 13081-18-0
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR